CD™ Vials with PTFE Lined Screw Cap, Chemglass
Supplier: Chemglass
CG-4915-01
89029-470EA 277.9 USD
89029-470
CD™ Vials with PTFE Lined Screw Cap, Chemglass
Vials
These CD™ vials have a unique patented conical interior bottom permitting the contents to completely drain to the center bottom of the vial.
  • Less than 4 µl of dead volume
Maximum extraction can be accomplished using a syringe or micropipette.
Delivery information: Available complete with open top screw caps having a PTFE faced silicone septa or vials only.