Ellipso-Spoon® Spatula, Chemglass
Supplier: Chemglass
CG-1987-04
89028-940EA 226.13 USD
89028-940
Ellipso-Spoon® Spatula, Chemglass
Spatulas
The specialty curved shape permits contact with all corners of bottles, flasks, and containers.
  • Made of polished stainless steel
  • Handle diameter is 8 mm
Crystals, powders, solid substances and thick liquids are easily retrieved and do not fall back into the container. The sampler has a spoon and spatula.