OptiChem® Heating Mantle Controller, Double Circuit, Variable Output Voltage, Chemglass
Supplier: Chemglass
CG-15010-20
89028-528EA 957.83 USD
89028-528
OptiChem® Heating Mantle Controller, Double Circuit, Variable Output Voltage, Chemglass
Heaters Heating Mantles
The heating mantle is similar to 10170-438, but the voltage output can be adjusted between 30 and 60 V.
  • Controller for use with CG-14000
  • Controller for use with CG-14002
  • Heating Mantles rated at 30 or 60 Volts
Delivery information: All units are supplied with a 6' separable power cord, a grounded, 3-wire receptacle and lighted power switch.