Beakers, Jacketed, Low Form, Chemglass
Supplier: Chemglass
CG-1107-12
89028-336EA 1501.82 USD
89028-336
Beakers, Jacketed, Low Form, Chemglass
Beakers
These beakers have a larger opening at the top of the jacket and a shorter overall height.
  • Width to height ratio on all sizes is 2:1
  • With a flat bottom on both the inner and outer vessel
  • Hose connections have an O.D. of 10 mm at the largest serration