Moisture Test Adapters, Chemglass
Supplier: Chemglass
CG-1063-01 CG-1063-04 CG-1063-05 CG-1063-09
89028-284EA 304.65 USD
89028-284 89028-290 89028-292 89028-300
Moisture Test Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters are used to construct a simple moisture trap.
Adapters with larger center to center spacing can be used with process reactors and larger flasks up to 50 l. Add a condenser to the top outer joint and a graduated funnel to the lower inner joint.