Vacuum Gauges, Digital, Chemglass
Supplier: Chemglass
AF-0301-B-01
89028-168EA 2112.87 USD
89028-168
Vacuum Gauges, Digital, Chemglass
Pumps Vacuum Gauges
Digital vacuum gauge has an analog style screen for easy viewing.
  • User selected units of measure
The transducer is made of corrosion resistant ceramic for durability and measures absolute pressures. Powered by a 9 volt lithium battery with adjustable sleep timer for long battery life.
Delivery information: Supplied with battery.