Whatman™ Reaction Folders - Sealable Hybridization Bags, Whatman products (Cytiva)
Supplier: Cytiva
10483064
89027-004PK 174.06 USD
89027-004
Whatman™ Reaction Folders - Sealable Hybridization Bags, Whatman products (Cytiva)
Bags Roll Stock
Whatman reaction folders are sealable bags for hybridization and incubation reactions.
  • Folders can be sealed with a standard heat-sealer
The folders are open on three sides to allow easier insertion of a wet membrane when compared to standard hybridization bags.