Glove Box Holders, Acrylic, IBI Scientific
Supplier: IBI Scientific
GD-10 GD-20 GD-30
89026-628EA 101.33 USD
89026-628 89026-630 89026-632
Glove Box Holders, Acrylic, IBI Scientific
Glove Dispensers and Glove Box Holders
Dispensers hold 1–3 standard boxes of gloves and help organize the laboratory
Constructed of sturdy 6.4mm (1/4") cast acrylic. Mounts vertically or horizontally on a wall. Built-in base allows it to stand upright on a countertop.
Ordering information: Supplied with mounting screws.