11 mm Standard Opening Crimp-Top Vials, Thermo Scientific
Supplier: Thermo Scientific
CERT4011-89W
66011-240CS 488.21 USD
66011-240 66030-248 82031-698 89026-588 66030-242 66030-244 66065-128
11 mm Standard Opening Crimp-Top Vials, Thermo Scientific
Vials
Crimp-top vials have a narrow neck compatible with 11 mm aluminum seals and 5 mm inserts.
Inserts are made of clear, borosilicate glass. These inserts are custom fitted for use with standard opening, 2 ml, 12×32 mm vials. Flat-bottom construction optimizes the depth of the vial.
The conical cavity allows maximum retrieval of contents with a syringe. All vials are manufactured from borosilicate glass, except for Glastic® plastic vials, which have a fixed glass 150 µl insert. Glass MicroVial™ vials are for limited volume applications.
Certifications: Certified vial kits are tested and certified to meet or exceed up to 17 critical performance parameters; certificate enclosed with kit.