VWR® Flathead [ST] Stoppers,PTFE
Supplier: VWR International
89026-340
89026-340EA 96.1 USD
89026-340
VWR® Flathead [ST] Stoppers,PTFE
Stoppers PE/LDPE/HDPE Stoppers
Inert and chemically resistant solid PTFE stoppers for use with standard ground glass joints.
  • Chemically resistant
  • Inert
  • PTFE fabrication
Ideal for sealing glass containers and glass jointware assemblies. Trapezoidal sealing rings provide high vacuum performance and reduce seizure, while the non-stick properties ensure easy removal. Usable to 280°C (536°F).