VWR® Scoop, PTFE
Supplier: VWR International
041.001
89026-220EA 117.1 USD
89026-220
VWR® Scoop, PTFE
Scoops
Non-contaminating PTFE scoop useful for transferring frozen substances and other non-liquids.
  • Chemically inert
  • Non-contaminating
  • PTFE construction
Scoop is suitable for use with frozen substances and other solids.