VWR® Ladle, PTFE
Supplier: VWR International
89026-192
89026-192EA 129.74 USD
89026-192
VWR® Ladle, PTFE
Samplers Sample Dippers
This PTFE molded sampling ladle is useful for transferring hot or cold reactive substances.
  • Chemically resistant
  • Inert
  • PTFE fabrication