Portable Gas Hot Plate, Global Market Connections
Supplier: Global Market Connections
HP-2003
89025-956EA 20232.57 USD
89025-956
Portable Gas Hot Plate, Global Market Connections
Heaters Hot Plates
This gas operated hot plate provides a fully adjustable infrared heat source of up to 1100°C (2000°F) without the need for an electrical outlet
Push-button auto-ignition makes lighting easy. The self-contained gas chamber refills quickly and easily with standard butane fuel. An optional beaker stand, constructed of high-quality black powder-coating on iron wire components supports beaker sizes of up to 1000mL while resisting corrosion and discoloration. The optional crucible cradle allows safe heating, cooling, and weighing of the crucible and its contents.