Precision Suture Scissors, OR Grade, Sklar
Supplier: Sklar
22-2822
89025-316EA 178.75 USD
89025-316
Precision Suture Scissors, OR Grade, Sklar
Scissors
Precision Suture Scissors are used for suture removal
This instrument has sharp tips, that easily slide under and gently lift sutures in preparation for removal. One of the sharp/sharp blades has a notch that the suture rests in before it is cut.