Potts-Smith TC Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
51-3460 51-3480
89025-100EA 278.64 USD
89025-100 89025-102
Potts-Smith TC Dressing Forceps, OR Grade, Sklar
Forceps
Sklar® Potts-Smith TC Dressing Forceps are most commonly used for application and changing of dressings. They may also be used for dissection of fine or single-layer tissue.
Cross-serrated tips provide a solid grip on both materials and tissue
Censitrac Ready products are industry standard and constructed of stainless steel.
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.