Ground Cord, Male Stud, SCS, Desco Industries
Supplier: Desco
CGC151M
89024-538EA 12.65 USD
89024-538
Ground Cord, Male Stud, SCS, Desco Industries
Grounders Grounding Wires
This ground cord connects two wrist straps and a worksurface to ground.
  • Compatible with mats that have a 10 mm socket snap
  • The 10 ring terminal at end of ground cord allows easy attachment to electrical ground
  • No resistor is required, but with resistor is acceptable
Delivery information: Includes 1 megohm resistor, meets Grounding standard ANSI/ESD S6.1.