Non-Marking Blue Heel Grounder, SCS, Desco Industries
Supplier: Desco
NS-HGC1M
89024-520EA 19.41 USD
89024-520
Non-Marking Blue Heel Grounder, SCS, Desco Industries
Grounders Heel and Toe Grounders
The blue sole of the Non-Marking Heel Grounder eliminates scuffs and marks on ESD floors.
  • Re-positionable configuration extends service life
  • Elastic loop provides a secure and comfortable fit
This quality grounder is tested for electrical performance and is lot numbered.
Certifications: ESD-SP9.2, ESD TR53. UL listed.