Economy Single-Wire Adjustable Wrist Strap, SCS, Desco Industries
Supplier: Desco
ECWS61M-1
89024-496EA 8.9 USD
89024-496
Economy Single-Wire Adjustable Wrist Strap, SCS, Desco Industries
Grounders Wrist Grounders
The Economy Single-Wire Adjustable Wrist Strap set provides a cost-effective means of grounding production employees.
  • One size fits all adjustable wrist band with 6' coil cord
  • Blue band, black coil cord
  • 1 megohm resistor
This wrist strap features an adjustable fabric wristband with alligator clip, and a 4 mm snap.
Certifications: Meets ANSI/ESD S20.20 and ANSI/ESD S1.1.