Surgi-OR™ Spencer Stitch Scissors, OR Grade, Sklar
Supplier: Sklar
95-346
89023-588EA 49.1 USD
89023-588
Surgi-OR™ Spencer Stitch Scissors, OR Grade, Sklar
Scissors
Surgi-OR™ Spencer Stitch scissors are used for suture removal.
They have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They have smaller, more delicate tips used for removing smaller sutures. They should not be used for cutting tissue.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.