Surgi-OR™ Knowles Bandage Scissors, Physician Grade, Sklar
Supplier: Sklar
95-254
89023-534EA 56.87 USD
89023-534
Surgi-OR™ Knowles Bandage Scissors, Physician Grade, Sklar
Scissors
Surgi-OR™ Knowles Bandage Scissors are durable, lightweight, and designed for cutting thick dressing material.
Bandage scissors have a blunt tip on the upper blade so as not to cause any injury to the patient while removing bandages or dressings.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
Surgi-OR™ instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with smooth, sharp/blunt tips, and a length of 5-1/2".