ESD Shallow Textured Bottom Tray, MFG Tray
Supplier: Molded Fiber Glass Tray
6290005167
89022-940EA 109.2 USD
89022-940
ESD Shallow Textured Bottom Tray, MFG Tray
Trays/Dishes/Pots Laboratory Trays/Dishes
Used in the assembly of circuit boards, computers, medical device, and other types of electronic or static sensitive assembly
Trays are designed for component assembly and for compatability with conveyors or cart transport systems. These trays feature a textured bottom for improved contact with conveyors.