NitriGuard Unsupported Nitrile Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
LA142G/7
89022-086CS 1138.08 USD
89022-086 89022-092 89022-094 89022-076
NitriGuard Unsupported Nitrile Gloves, Honeywell Safety
Gloves
Ideal for chemical handling and processing, food processing, aerospace and automotive degreasing, automotive assembling and painting, and graphic arts and print shop cleanup.
  • Soft flock lining absorbs perspiration and eases donning and doffing
  • Curve-shaped hand design provides a natural fit
  • Latex-free
The 100% nitrile content offers superior resistance to cuts, snags, abrasions, and punctures. Gloves provide excellent perfomance when exposed to solvents, caustics, and animal fats. Gloves feature a sanitized interior with sandpatch grip and straight cuff.
Certifications: Gloves comply with USDA and FDA regulations 21CFR for use in food processing.