Diamond-Plate Military Switchboard Anti-Fatigue Matting, Wearwell®
Supplier: Wearwell
712.316X3X75GY
89021-402RL 3713.7 USD
89021-402
Diamond-Plate Military Switchboard Anti-Fatigue Matting, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
Military Switchboard matting prevents life-threatening electrical shocks in high-voltage environments.
  • Meets Mil Spec 15562-G Amend. 3, Type III
  • Dielectric Strength: 30,000 volts
This non-conductive matting delivers a dielectric strength of 30,000 volts. The easy to clean surface offers exceptional ozone and oil resistance. Ideally suited for use in dry environments.
Certifications: Meets ASTM F1677. Dry: COF = 0.68. Passes Mil. 15562-F and FMVSS 302.