Rubber Pipette Fillers
Supplier: Heathrow Scientific
HS20632B
89015-588CS 4734.32 USD
89015-588
Rubber Pipette Fillers
Pipettes Pipette Bulbs
Rubber safety pipette filler is designed to facilitate one-handed operation.
  • Capacity: 35 ml
  • Contoured shape and heavy wall neck increase grip for easier use with pipettes and syringe
  • Made of natural rubber
  • Control air evacuation, liquid uptake, and liquid dispensing with three valves
  • Pinch release valve for smooth liquid flow
  • Filler fits standard size pipettes
Red pipette filler for safe pipetting of caustics, it does not contain carbon black and will not smudge.
Dimensions (L×O.D): 130×52 mm
Hole diameter: 7 mm