Whizard® Defender 7 Extended Cuff Gloves, Wells Lamont
Supplier: Wells Lamont
135437 135438 135439 135440 135441
89015-531EA 37.32 USD
89015-531 89015-533 89015-535 89015-537 89015-539
Whizard® Defender 7 Extended Cuff Gloves, Wells Lamont
Gloves
The prewashed and preshrunk gloves feature Honeywell Spectra® Fiber.
  • Spectra fiber
  • Antimicrobial fibers
The combination of durable fibers and stainless steel provides excellent tensile strength.
The ambidextrous gloves work best in heavy-duty applications.
Antimicrobial fibers offer an additional layer of protection by inhibiting bacterial and fungal growth. Superior cut resistance eliminates the high cost associated with lacerations from sharp-edged machinery and hazardous applications and environments.
Certifications: ANSI Cut Level 5, A7. CFR 21 approved for food contact.
Ordering information: The gloves are sold in multiples of six pieces.
Caution: Caution Information: Cut-resistant gloves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades.