SP Bel-Art Micro-Spoon and Spatula Sampling Set, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36705-0000
89013-642PK 148.83 USD
89013-642
SP Bel-Art Micro-Spoon and Spatula Sampling Set, Bel-Art Products, a part of SP
Spoons
Each tool has two different end shapes to better match user needs.
  • Polished stainless steel construction
  • Great variety of tools for sampling
  • Durable for a long life and easy cleaning
  • Autoclavable
Tool ends include two small spoons for transferring and collecting both dry and liquid material, and eight flat spatulas of various widths and lengths for mixing, scraping, and spreading materials.
Six of the tools have square ends and two are rounded for working in test tubes. Two V-shaped spatulas of different sizes can be used for lifting samples or spill-free transferring of materials. The blunt-tip tweezers feature serrated inside surfaces for securely handling samples and grooved handles for a better grip.
Kit includes six different small double-ended tools and a set of tweezers. All tools are constructed of polished stainless steel for long life and easy cleaning. A carrying case protects and organizes the tools when not in use. Tools are steam autoclavable at 121°C (250°F).