SAF-T-FIT™ Plus P1130 Respirators, Honeywell Safety
Supplier: Honeywell Safety Products
14110439
89013-637BX 101.11 USD
89013-637 89013-638
SAF-T-FIT™ Plus P1130 Respirators, Honeywell Safety
Respirators Disposable Respirators Particulate Respirators
SAF-T-FIT Plus molded cup disposable respirators offers variety in protection, size and comfort.
  • Nose clip is color-coded for visible size recognition
  • Latex free and durable suspension straps
  • Low breathing resistance
  • Humidity-resistant filter media