VWR® Foam Wall Mop Head
Supplier: VWR International
89012-764
89012-764CS 432.33 USD
89012-764
VWR® Foam Wall Mop Head
Mops
Foam wall mop heads are developed for cleaning ceilings and walls in controlled and critical environments.
  • Autoclavable
  • Free of any cellulosic contaminants
Compatible with a wide range of chemicals, making it ideal for the application of disinfectants and cleaning solvents
Suitable for ISO Class 5 (FED-STD-209E Class 100/M3.5) environments.
Made of polyester fabric laminated to clean ester foam. Compatible with gamma, ETO, and autoclave sterilization. Fits a 15.2 cm (6") frame. Dimensions: 17.8Lx10.4Wx3.2Hcm (7x4x11/4").
Ordering information: Mop head can be used with mop head frame 33503-126 and telescoping mop handles (see 68300-353 series and 68310-408 series). A small downpress wringer insert is required to reduce the size of the downpress wringer cavity for proper compression of the mop head. Contact your Avantor sales representative for more information.