RC2 Rodent Circuit Controller Anesthesia System, VetEquip®
Supplier: Vetequip
922100
89012-606EA 7993.59 USD
89012-606
RC2 Rodent Circuit Controller Anesthesia System, VetEquip®
Animal Supplies Animal Anesthesia Systems
This anesthesia system maintains an accurate flow rate to any circuit, unlike stopcock or gang-valve delivery systems.
  • Short, easy-to-follow protocol requires no calculations and no flowmeter adjustments
  • Six individual connections allow any combination of connections to breathing circuits and chambers
  • Specific gas flow is provided to each animal, regardless of the number of circuits or the length of the tubing
  • Lower preset flows create less waste gases, resulting in lower trace exposure to the user
The system can be placed on a countertop, mounted on a wall, or installed on a cart to make the system mobile.
Ordering information: The complete anesthesia system is supplied with a manufacturer’s one-year money-back guarantee and seven-year replacement warranty; “VetEquip Anesthesia Book” and DVD; isoflurane funnel-fill vaporizer; anti-spill adapter for vaporizer; 0 - 4 L/min. O2 flowmeter; O2 flush valve; supply pressure gauge; system pressure gauge; two 1 L (0.26 gal.) induction chambers; button-operated flush assemblies for both chamber circuits; four Mapleson E breathing circuits; four 12 mm (¹⁵⁄₃₂"") nose cones; 3 m (10') O2 hose with DISS fittings; 1.8 m (6') of 22 mm (⅞") evacuation tubing; six charcoal filters; and T-shirt. Each Mapleson E circuit comes with 6mm (¼") fresh gas tube within a 22 mm (⅞") waste gas tube. Non-adjustable O2 regulators (89012-466 and 89012-468) are preset to 50psi and have DISS male connections. For mobile option or for configurations with other vaporizers, induction chambers, and circuits, contact your VWR sales representative.