IMPAC6 Integrated Multi-Patient Anesthesia Center, VetEquip®
Supplier: Vetequip
901808
89012-496EA 8552 USD
89012-496
IMPAC6 Integrated Multi-Patient Anesthesia Center, VetEquip®
Animal Supplies Animal Anesthesia Systems
This anesthesia center is ideal for high-volume, assembly-line type procedures
Anesthesia can be induced and maintained in four chambers. Two Bain breathing/procedure circuits, one on either side of the machine, allow two people to perform procedures simultaneously for increased productivity. The built-in CO2 chamber circuit can be used for O2 recovery, O2/CO2 short-term anesthesia, or euthanasia. The induction chambers have positive sealing lid gaskets to contain anesthetic gases. Gas flow to the chambers and breathing circuits can be individually controlled. The nose cones supplied with the system have virtually no dead space, which helps eliminate CO2 buildup that can result in death under anesthesia. The internal evacuation manifold collects waste gases from all the circuits and channels them to the back of the system via a single outlet. The precision, funnel-fill isoflurane vaporizer features temperature and flow rate compensation. The aluminum unit has a baked enamel finish.
Ordering information: The complete anesthesia center is supplied with a manufacturer’s one-year money-back guarantee and seven-year replacement warranty; “VetEquip Anesthesia Book” and DVD; isoflurane funnel-fill vaporizer; anti-spill adapter for vaporizer; four induction chambers; separate CO2 chamber with stainless steel shelf; two breathing circuits; flowmeters for O2 and CO2; line pressure gauges for O2 and CO2; two 9 mm (¹¹⁄₃₂"") mouse-size nose cones; four 14 mm (⁹⁄₁₆") medium/large rat size nose cones; 3 m (10') O2 hose with DISS fittings; 3 m (10') CO2 hose with DISS fittings; 1.8 m (6') of 22 mm (⅞") evacuation tubing; waste gas evacuation port; charcoal filters; and T-shirt. Divided chamber 89012-700 has two W×D×H 108×254×102 mm (4¼×10×4") compartments. Non-adjustable gas-specific regulators are preset to 50psi and have DISS male connections.