Mini Cable Lockout, Brady®
Supplier: Brady Worldwide
50940
89011-766EA 115.98 USD
89011-766
Mini Cable Lockout, Brady®
Security Locks
Compact and portable lockout made of impact-modified nylon providing superior chemical, corrosion, and temperature resistance.
  • Perfect for use on disconnect switches and small gate valves
  • Six lock holes accommodate most lock shackles and hasps
  • Lockout features front legend that reads: “DANGER, LOCKED OUT, DO NOT REMOVE”
Includes 2.4m (8') nonconductive, vinyl-coated steel cable, 1.8cm (11/16") in diameter. Cable is stored internally with a push-button, self-winding mechanism that automatically cinches it tight.