VWR® Foam-Tipped Swab
Supplier: VWR International
89009-338
89009-338CS 4335.41 USD
89009-338
VWR® Foam-Tipped Swab
Swabs Swabs for Sampling
Foam tip is made from soft and pliable medical-grade 100ppi polyurethane foam.
  • Ideal for critical environments
  • Polystyrene shaft
  • Soft and pliable tip
The fine-pore, high-absorbency tip is specifically designed to retain and transfer fluids. Polystyrene shaft is scored 74.4mm (2.93") from tip.