SP Bel-Art Secador® Auto-Desiccator Module Desiccant Replacement Beads, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F42074-0020
89009-210EA 22.77 USD
89009-210
SP Bel-Art Secador® Auto-Desiccator Module Desiccant Replacement Beads, Bel-Art Products, a part of SP
Desiccators Desiccants
Color-indicating silica gel beads for use in refilling the Secador® auto-desiccator module.
  • Color-changing moisture indicator beads
  • Inert, non-toxic, and non-corrosive
  • Replacement beads indicate moisture saturation and replacement time
Silica gel absorbs moisture from the air and is inert, non-toxic and non-corrosive
Beads contain a color-changing moisture indicator that turns from blue to light pink, indicating that the silica gel is saturated and can not absorb additional moisture.
Secador® auto-desiccator modules automatically regenerate the silica gel beads for thousands of cycles; however, when beads turn pink/dark brown, and fail to regain their blue color, it is time to replace the beads with a fresh supply.
Unlike other desiccants, silica gel is non-friable and won’t disintegrate into powder even after repeated regeneration.