SP Bel-Art Standards Bottle Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18513-2300
89009-198EA 159.37 USD
89009-198
SP Bel-Art Standards Bottle Rack, Bel-Art Products, a part of SP
Racks Bottle Racks
This clear acrylic rack organizes valuable USP™ reference standards while allowing easy viewing of bottle labels.
  • Clear acrylic construction
  • Easy viewing of bottle labels
  • Suitable for low humidity storage
  • Compact design
Use in Secador® 1.0–4.0 desiccators for low humidity storage. Rack stores thirty 23 mm standards bottles in a compact footprint.