Digital 'Pocket' Cutting Oil Refractometer, Model PAL-102S, ATAGO®
Supplier: Atago
4502
89008-814EA 830.22 USD
89008-814
Digital 'Pocket' Cutting Oil Refractometer, Model PAL-102S, ATAGO®
Refractometers Handheld Refractometers
Designed for controlling the concentration of cutting oils
The device features IP65-rated dustproof and water-resistant protection, and Automatic Temperature Compensation for accurate measurements in shifting environmental conditions. A sample volume of 0.3mL is required for measurement results.
Molding lubricants and rust resistance liquids are also within the measurement range of this device. Refractometer is best suited for determining the percent dilution and controlling the percent concentration of hydraulic oil and cleaning liquids.
Ordering information: Uses two AAA batteries.