Digital 'Pocket' Refractometer, Model PAL-[alpha], ATAGO®
Supplier: Atago
3840
89008-788EA 915.91 USD
89008-788
Digital 'Pocket' Refractometer, Model PAL-[alpha], ATAGO®
Refractometers Handheld Refractometers
Economic pocket refractometer offers a wide Brix range
It is suitable for measuring most sample types, especially those involved in cooking processes. Refractometer features IP65-rated dustproof and water-resistant protection. The unit can be washed under running water for simplified cleaning. Equipped with Automatic Temperature Compensation to ensure accurate measurements, even in shifting environmental conditions. Accepts sample volumes of 0.3mL.
Ordering information: Uses two AAA batteries.