Large Universal Valve Lockout Base Clamping Unit, Brady Worldwide®
Supplier: Brady Worldwide
50899
89007-220EA 124.1 USD
89007-220
Large Universal Valve Lockout Base Clamping Unit, Brady Worldwide®
Clamps General Purpose Clamps
Universal valve lockouts are effective for all types of standard valves
  • Versatile and rugged
  • lightweight and easy-to-use
New open-ended clamp fits over closed rings and wide handles. Constructed of industrial-grade steel and nylon for extra impact and chemical resistance.
They can also be used to lock out large levers, T-handles, and hard-to-secure mechanical devices.
Ordering information: Cable and blocking arm sold separately.