E-A-R™ Classic™ SuperFit™ Earplugs, 3M™
Supplier: 3M
7000002333
89006-804BX 132.17 USD
89006-804 89006-822 89006-772 89006-820 89006-840
E-A-R™ Classic™ SuperFit™ Earplugs, 3M™
Earplugs Disposable Earplugs
3M™ E-A-R Classic™ SuperFit™ Earplugs feature an orange fitting ring that acts as a built-in visual check.
If the ring is visible after insertion, refitting is recommended for best hearing protection.