Axygen® Macrovolume Pipette Tips
Supplier: Corning
T-10ML-C
89005-641CS 1135.83 USD
89005-641
Axygen® Macrovolume Pipette Tips
Pipette Tips
These tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most brands of pipettors.
  • These polypropylene tips are RNase-/DNase-free
  • Nonpyrogenic
  • Compatible with Gilson P10 ml and MLA pipettors
  • Compatible with Eppendorf® pipettors
  • Eppendorf® is a registered trademark of Eppendorf SE
They are available in Maxymum Recovery, beveled, non sterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.