Axygen® Universal Macro Volume Pipette Tips, Corning
Supplier: Corning
T-5000-C
89005-638CS 626.61 USD
89005-638 89005-640
Axygen® Universal Macro Volume Pipette Tips, Corning
Pipette Tips
Axygen® Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors.
  • Polypropylene tips are RNase-/DNase-free
  • Non pyrogenic
  • Graduation marks at 1, 2, 3, 4, 5 mL and 0.5 mL increments in between
  • Compatibility: Gilson P5000 and MLA pipettors
  • Compatible with Eppendorf® pipettors
Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.