Wet-Strengthened Qualitative Filter Paper, Grade 992, Ahlstrom
Supplier: Ahlstrom
9920-3400
89005-524PK 165.78 USD
89005-524
Wet-Strengthened Qualitative Filter Paper, Grade 992, Ahlstrom
Filters Filter Paper Qualitative Filter Paper
Ahlstrom 992 is a fast filtering grade with a creped surface and high loading capacity.