VWR® Benchtop PCR Cryogenic Rack
Supplier: VWR International
89004-570
89004-570CS 894.93 USD
89004-570
VWR® Benchtop PCR Cryogenic Rack
Coolers Benchtop Coolers
Benchtop rack allows frozen PCR samples to be worked with for up to four hours at 0°C (32°F).
  • Refreezable rack holds 0.2 mL tubes, strips, and plates
  • As the rack warms, the color of the chilling surface changes from purple to pink to indicate when the samples need to be returned to the freezer
Rack is constructed of polypropylene and polygel and includes a snap-on lid.