Shell Vials, Borosilicate Glass, with Plug-Style Needle Closure, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
60831D-830 60831D-843 60831D-1544 60835D-1544 60831D-1231
66009-902CS 728.2 USD
66009-902 66009-924 66009-968 66009-990 89003-428
Shell Vials, Borosilicate Glass, with Plug-Style Needle Closure, Kimble Chase
Vials
TITESEAL® shell vials are ideal for chromatography applications.
  • White polyethylene plug style needle closures, unattached
  • Ideal for use with light-sensitive samples
  • Made of 51 expansion borosilicate amber glass
Certifications: Meets USP Type I and ASTM E438, Type I, Class B requirements