KIMAX® Centrifuge Tubes, Conical-Bottom, with Flathead PTFE Stoppers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
45174-13
89002-204CS 286.22 USD
89002-204 21021-420
KIMAX® Centrifuge Tubes, Conical-Bottom, with Flathead PTFE Stoppers, Kimble Chase
Tubes Centrifuge Tubes
Standard Taper KIMAX® tube is made with heavier-walled tubing to withstand higher centrifugation speeds.
  • Manufactured from 33 expansion, low extractable borosilicate glass
  • Conical bottom
  • Color-coded flathead PTFE stopper
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.