Graduated Dairyware Centrifuge Tube, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
45167-50
89002-202CS 605.62 USD
89002-202
Graduated Dairyware Centrifuge Tube, Kimble Chase
Tubes Centrifuge Tubes
Designed for use in the determination of the solubility index of dry milk solids or whey products as specified by the American Dairy Products Institute.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Also used in pulp determination of citrus products. The top is beaded for strength and all markings are durable black ceramic enamel. Graduated and calibrated to contain.