KIMAX® Dean-Stark Distilling Receiver with [ST] Joints, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
22012-10
89001-750CS 381.32 USD
89001-750
KIMAX® Dean-Stark Distilling Receiver with [ST] Joints, Kimble Chase
Distillation Components Distillation Receivers
Modified distilling receiver with [ST] 24/40 joints, siphon return arm, and a sidearm bridge that equalizes pressure.
Designed from ASTM Specification E123, Style A.