KIMAX® Powder Funnels, Kimble Chase, DWK Life Sciencese
Supplier: DWK Life Sciences (KIMBLE)
29020-60 29020-80 29020-100 29020-125 29020-150
89001-416CS 567 USD
89001-416 89001-418 89001-420 89001-422 89001-424
KIMAX® Powder Funnels, Kimble Chase, DWK Life Sciencese
Funnels Powder Funnels
Funnels feature short, wide stems and fire-polished tops and stems.
These powder funnels are made of glass.
Certifications: Designed from ASTM specification E1095, Type 4 requirements.