KIMBLE® KIMAX® Gooch Crucibles, High Form, with KIMFLOW® Fritted Disc, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
28250-302
89001-364CS 63.98 USD
89001-364
KIMBLE® KIMAX® Gooch Crucibles, High Form, with KIMFLOW® Fritted Disc, DWK Life Sciences
Crucibles
Suitable for precipitates to be dried to constant weight at 110°C
Note: Exposure to sudden temperature changes should be avoided.
For higher temperatures, an electric furnace is advised and crucibles should not be removed until temperature has dropped to 250°C