KIMBLE® RAY-SORB® Amber Flat Bottom Short Neck Boiling Flask, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
25057-250
89001-270CS 141.46 USD
89001-270
KIMBLE® RAY-SORB® Amber Flat Bottom Short Neck Boiling Flask, DWK Life Sciences
Flasks Round Bottom Flasks
Full-length, short neck flasks are RAY-SORB® processed to provide protection to solutions sensitive to short-wavelength light.
Designed from ASTM Specification E 1403, Type I, Class 4 requirements.