KIMAX® Single Metric Scale Graduated Cylinder, Emulsion Test, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
20011-100
89001-092CS 68.31 USD
89001-092
KIMAX® Single Metric Scale Graduated Cylinder, Emulsion Test, Kimble Chase
Cylinders Measuring Cylinders
Used to determine emulsifying and demulsifying tendencies of lubricating oils (ASTM D1401)
Round base is designed to fit baths in which the cylinder is generally used. Calibrated to contain with a durable white ceramic enamel scale.